miniPCR® Lab in a Box™ Kit #2
Fournisseur: AMPLYUS LLC
Certificats
À propos de cet article
Your complete set up, ready to go!
All the hardware you need to get your molecular biology lab up and running. And you can take it anywhere!
Delivery: The miniPCR® Lab in a Box™ Kit #2 includes: miniPCR® mini8X or mini16X thermal cycler: A powerful, portable PCR machine always at your fingertips; blueGel™ system: All-in-one electrophoresis and transillumination; Set of three micropipettes (1 to 10 μl, 2 to 20 μl, 20 to 200 μl): Universal fit, compatible with standard micropipette tips, and easy to re-calibrate; Fieldable carrying case with shoulder strap, semi-rigid shell and protective foam inserts; 200 μl micropipette tips (2 racks of 96), and 10 μl micropipette tips (2 racks of 96).
Frequently Bought Together