ContactUsLinkComponent
Hydrogen and High Purity Air Generator for GC-FID
Catalogue # 76836-908
Fournisseur:  LUCIDITY SYSTEMS LLC
undefined
Hydrogen and High Purity Air Generator for GC-FID
Catalogue # 76836-908
Fournisseur:  LUCIDITY SYSTEMS LLC
Supplier Number:  928276
Spécifications
  • Description:
    Hydrogen and high purity air generator for the Lucidity GC-FID, 10.5×18×12.5"
  • Cat. no.:
    76836-908
Spécifications
À propos de cet article
Generator that produces all necessary gasses required to run the Lucidity GC-FID.
  • Hydrogen and high purity air generator for the Lucidity GC-FID
  • Small footprint and economical
  • Self-installable instrument that only requries DI-H₂O
The Lucidity GAS produces hydrogen and high purity air. The economical GAS with its small footprint compliments the Lucidity GC-FID perfectly. The GAS produces all necessary gasses required to run the Lucidity GC-FID. Just add DI-H2O to the self-installable instrument and your lab can have access to gasses required for analysis immediately. The Lucidity GC-FID and GAS communicate with each other, making the perfect and safest pair of instrumentation. With a 24-month warranty the GAS is a great solution for all labs.