- Magnification:4×, 10×, 40×
- Description:Student monocular microscope with disc diaphragm
- Observation method:Brightfield
- Illumination:LED
- Stage:Standard
- Type:Monocular
- Cat. no.:470351-240
Designed to provide educators with a student-ready instrument at an affordable price.
- DIN 4×, 10×, 40×R objectives
- Flat top stage with stage clips and disc diaphragm
- Coarse and fine focusing control
- Variable LED illuminator
- 3500 mah NI rechargeable battery
A robust microscope that is ideal for the educational setting: compact footprint, built-in LED illumination, built-in carry handle, simple set-up and storage. Designed for elementary and middle school application, the EXM-50 offers DIN Achromat 4×, 10×, 40×R* objectives (* spring loaded).
The EXM-50 features an LED illumination system allowing corded and cordless operation. For cordless operation, simply insert AA alkaline batteries (not included) into the battery compartment on the bottom of the microscope. The EXM-50 is super portable thanks to its integrated carrying handle making it easier than ever for students and teachers to move the microscope around the classroom. The viewing head is rotatable 360° making the EXM-50 easy to store, too.