Long Fragment Buffer Expansion | Avantor
Long Fragment Buffer Expansion
LFB expansion, All flow cells, 4
Catalog # 76848-064
Fournisseur #:  EXP-LFB001
Long Fragment Buffer Expansion
Catalog # 76848-064
CAS Number:  
Long Fragment Buffer Expansion
Long Fragment Buffer Expansion
LFB expansion, All flow cells, 4
CAS Number:  
Price
$51.80
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
Additional Long Fragment Buffer (LFB).
The LFB Expansion contains additional Long Fragment Buffer (LFB) for use with sequencing experiments where shorter fragments (<3 kb) are to be removed. The LFB Expansion contains enough reagents for four reactions.
Delivery information: Kits are shipped at 2 to 8 °C in an environmentally-friendly temperature-controlled shipping box. Upon receipt, please place the product in a long-term storage location at −20 °C. Oxford Nanopore Technologies deem the useful life of the product to be 3 months from receipt by the customer.
Spécifications
Spécifications
  • Désignation:
    LFB expansion
  • Compatible with:
    All flow cells
  • No. of Reactions:
    4