ContactUsLinkComponent
Short Fragment Eliminator Expansion
  76848-058
 :  
undefined
Short Fragment Eliminator Expansion
  76848-058
 :  EXP-SFE001
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Short fragment eliminator expansion
  • Compatible with:
    All flow cells
  • Cat. no.:
    76848-058
  • No. of Reactions:
    30
An expansion pack designed for size selection of high molecular weight DNA samples to progressively deplete short fragments of <25 kb.
  • Size select to progressively deplete short fragments (<25 kb) prior to sequencing library preparation
  • Enrich sequencing libraries for long fragments without using any external third-party reagents
The Short Fragment Eliminator (EXP-SFE001) Expansion pack is recommended for users who want to size select high molecular-weight (HMW) gDNA to deplete short fragments (<25 kb) using size selective precipitation. The kit contains Short Fragment Eliminator (SFE) buffer which progressively removes fragments up to 25 kb, with fragments under 10 kb being almost completely removed in a quick and simple workflow.
Note: This kit is designed to remove short DNA fragments and requires samples to be predominantly made up of HMW DNA (>48 kb). If there are large amounts of short fragments in your sample, this will result in a low yield at the end of the prep and a low sequencing output.
 : Kits are shipped at 2 to 8 °C in an environmentally-friendly temperature-controlled shipping box. Upon receipt, please place the product in a long-term storage location at −20 °C. Oxford Nanopore Technologies deem the useful life of the product to be 3 months from receipt by the customer.