To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Rapid Adapter Auxiliary V14
- Compatible with:R10.4.1 flow cells
- Cat. no.:76848-060
- No. of Reactions:6
Spécifications
À propos de cet article
Additional Rapid Adapter and Adapter Buffer to enable customers to maximise the use of Rapid Barcoding Kits 24 and 96 V14 (SQK-RBK114.24 and SQK-RBK114.96).
The Rapid Adapter Auxiliary V14 (EXP-RAA114) Expansion contains additional Rapid Adapter and Adapter Buffer for another six reactions. This is to enable customers to use any unused barcodes in their Rapid Barcoding Kits V14.
To use this kit, ensure to purchase the Sequencing Auxiliary Vials V14 (EXP-AUX003) to provide the flow cell priming reagents.