ACCU-SCOPE® 3012 Upright Microscopes
Supplier: ACCU-SCOPE INC SE
Certificates
About this item
Economical, high-quality alternative for today’s budget-conscious world.
- Binocular, trinocular, or binocular ergonomic viewing head
- Head Inclined 30°, rotatable 360°
- WF10×/22 mm focusing eyepieces with eye guards
- Reversed quintuple nosepiece
- AIS E Plan Infinity 4×, 10×, 40×R and 100×R oil objectives
- Rackless mechanical stage with low position coaxial controls
The 3012 series features our E-Plan Achromat Infinity objectives which are renowned for crisp, high contrast images with outstanding resolution. With 22 mm field of view eyepieces and an eyepoint height that is adjustable, it can be raised 35 mm by rotating the eyepiece tubes up 180° to accommodate the needs of various users. The modular design of the 3012 allows options for entry level fluorescence applications, phase contrast, darkfield and simple polarization.
Frequently Bought Together