ACCU-SCOPE® EXC-350 Upright Microscopes
Supplier: ACCU-SCOPE INC SE
Certificates
About this item
Delivers a high level of optical performance, versatility and durability for clinical, veterinary and education laboratory environments.
- Trinocular viewing head
- Digital microscope options
- Infinity corrected
- Inclined 30°
- Optional fluorescence observation
- 3 W LED with Variable Intensity Control
- PL10×/20 mm focusable eyepiece with diopter for ease of use for the visually impaired
- Reversed quintuple nosepiece with space to add objectives
- Infinity corrected PLAN 4×, 10×, 40×R and 100×R oil objectives
- Mechanical stage with low position coaxial controls, right hand control
With our infinity plan objectives and brilliant LED illumination, the EXC-350 produces bright, crisp, and sharp images. The trinocular port is ready to accept a microscopy camera (requires c-mount camera adapter) for documentation, analysis, and instructional purposes. The EXC-350 is available with an optional 1, 2, or 3-channel fluorescence accessory with integrated LED illumination. Applications for fluorescence observation include immunostaining, fungus and bacteria identification, nucleic acids and genetics, and protein localization.
Frequently Bought Together