ACCU-SCOPE® EXC-120 Upright Microscopes
Supplier: ACCU-SCOPE INC SE
Certificates
About this item
The EXC-120 Series is engineered for classroom, medical and veterinary applications.
- Binocular, Monocular or Trinocular viewing head
- Digital microscope options
- Head is rotatable 360°
- WF10×/18 mm eyepiece (paired - one with pointer)
- Diopter adjustment for ease of use with the visually impaired
- Reversed quadruple nosepiece
- DIN 4×, 10×, SP40×R and 100×R oil objectives
- Rackless mechanical stage
Delivers sharp, high contrast objectives, ergonomic operation, and a rugged, die-cast aluminum alloy frame, the EXC-120 series will deliver excellent performance through years of heavy use. ACCU-SCOPE® offers the EXC-120 series with the options of brightfield observation, darkfield observation, phase contrast, simple polarization, in a trinocular, binocular, monocular or dual view observation viewing head, and the option of adding a camera for your digital microscope options.
Specifications
- Catalog No:
- 470351-162
- 470351-090
- 470351-174
- 470351-154
- 470351-156
- 470351-228
- 470351-076
- 470351-218
- 470351-142
- 470351-250
- 470351-164
- 470351-194
- 470351-258
- 470351-282
- 470351-086
- 470351-272
- 470351-264
- Observation method:
- Brightfield
- Brightfield
- Brightfield
- Brightfield
- Brightfield
- Brightfield
- Brightfield
- Brightfield
- Brightfield
- Brightfield
- Brightfield
- Brightfield
- Brightfield
- Brightfield
- Brightfield
- Brightfield
- Brightfield
- Stage:
- Rackless mechanical
- Rackless mechanical
- Rackless mechanical
- Rackless mechanical
- Rackless mechanical
- Rackless mechanical
- Rackless mechanical
- Rackless mechanical
- Rackless mechanical
- Rackless mechanical
- Rackless mechanical
- Rackless mechanical
- Rackless mechanical
- Rackless mechanical
- Rackless mechanical
- Rackless mechanical
- Rackless mechanical
- Illumination:
- LED
- LED
- LED
- LED
- LED
- LED
- LED
- LED
- LED
- LED
- LED
- LED
- LED
- LED
- LED
- LED
- LED