Setting a new standard for quality, versatility and performance in a compact stereo microscope.
- 1×, 2×, 3× and or 4× objective magnifications
- Binocular viewing head
- Inclined 45 degrees, rotatable 360 degrees
- WF10×/20 mm eyepieces with eye guards
- Frosted glass and black and white plates, dust cover
- Variable reflected and transmitted rechargeable LED illumination
EXS-210 series offers educators and industry an exceptional value at an affordable price. An economical stereo microscope with superior optical performance, the EXS-210 stereo microscope series is a student-proof microscope offering tremendous value for schools and hobbyists. With a built-in carrying handle and cordless operation, the EXS-210 series simplifies classroom set-up. Utilizing our state of the art optical coating techniques our student microscopes provide outstanding images at reasonable prices. The EXS-210 Series is available in 1×/2×, 1×/3×, 2×/4×, 1×/2×/3× and 1×/2×/4× configurations. Each microscope is equipped with LED illuminators for transmitted and incident illumination. The illuminators are designed to work separately or simultaneously. The LED illuminators are rechargeable for corded or cordless operation.
- Catalog No:
- 470351-244
- 470351-202
- 470351-214
- 470351-184
- Type:
- Binocular
- Binocular
- Binocular
- Binocular
- Observation method:
- Brightfield
- Brightfield
- Brightfield
- Brightfield