Estes Wizard Model Rockets | Avantor
Estes Wizard Model Rockets
Supplier:   Estes Industries
Estes Wizard Model Rockets
Supplier:  Estes Industries
Certificates
Estes Wizard Model Rockets
Estes Wizard Model Rockets
2 Options Available Below
Product Details & Documents
Intermediate-level rocket with laser-cut balsa fins and streamer recovery system.
  • Intermediate-level build
  • Requires standard size 18 mm A, B, or C engines (A or B recommended)
  • Max. altitude 488 m (1600')
Looking for a magical experience? Are your model rocketry skills ready to level-up? The Estes wizard rocket is exactly what you need! This high-performance rocket is easy to build and known for casting spells on all who own it.
With powerful Estes model rocket engines, this light-weight rocket can reach heights of up to 1600 feet in the blink of an eye. Its laser-cut balsa wood fins still make assembly easy, and its streamer recovery system helps it descend efficiently and safely from high altitudes. This sleek rocket is fun to build and thrilling to fly.
Ordering information: Launch system and rocket engines available separately.
Single kit includes all parts needed to assemble (1) model rocket. Some tools and glue may be required.
Bulk pack includes: (12) single kits.
Product Family Options
Informations sur le produit
  • Désignation
    Packaging
    Availability
    Price
  • Wizard model rocket kit (single)
    EA
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $13.55
    $13.55
    Liquidation
    Each
       $13.55
    Supplier Number: 1292
  • Wizard model rocket kit (bulk pack)
    Pack of 12
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $191.65
    $191.65
    Each
       $191.65
    Supplier Number: 1754