Estes Alpha Model Rockets
Estes Alpha Model Rockets
Supplier:   Estes Industries
Estes Alpha Model Rockets
Supplier:  Estes Industries
Certificates
Estes Alpha Model Rockets
Estes Alpha Model Rockets
2 Options Available Below
Product Details & Documents
Easy-to-assemble intermediate-level rocket with laser cut balsa fins.
  • Intermediate-level build
  • Requires standard size 18 mm A, B, or C engines
  • Max. altitude 305 m (1000')
The Estes alpha is a rocket that has stood the test of time and continues to be the go-to choice for educators after 40 years. Designed to be easy to build and customize, it offers a flexible platform for teaching valuable STEM principles to students. The unforgettable experience of launching an Estes alpha rocket is something that stays with people for a lifetime, with millions of individuals remembering their first launch with fondness.
With its easy assembly and reliable performance, the Alpha is sure to hook any student who gives it a try. Not only is Alpha easy to build and fly, it also looks great with its suggested color scheme and red, white, and blue decals. And with the ability to reach cloud-topping altitudes of 1000 feet, this rocket is sure to provide hours of entertainment.
Ordering information: Launch system and rocket engines available separately.
Single kit includes all parts needed to assemble (1) model rocket. Some tools and glue may be required.
Bulk pack includes: (12) single kits.
Product Family Options
Informations sur le produit
  • Désignation
    Packaging
    Availability
    Price
  • Alpha model rocket kit (single)
    EA
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $23.10
    $23.10
    Liquidation
    Each
       $23.10
    Supplier Number: 1225
  • Alpha model rocket kit (bulk pack)
    Pack of 12
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $308.75
    $308.75
    Liquidation
    Each
       $308.75
    Supplier Number: 1756