Autoclave Bags with Steam Indicator, Self-Seal, Heavy Duty SMS
Fournisseur: KEYSTONE ADJUSTABLE CAP CO., INC.
Certificats
À propos de cet article
Self Sealing Heavy Duty SMS Autoclave Bags from Keystone Cleanroom Products are designed to satisfy the most stringent requirements in the sterilization of critical pharmaceutical equipment and components.
- Convenient, time saving self-seal closure system
- Significant improvement versus cutting and wrapping with sheets and tape
- Superior barrier prevents liquid and microbial penetration
- Manufactured and packaged in a certified ISO 6 Cleanroom
- Strong, rip and tear resistant 50 gsm SMS material
- Sub packed into sealed labeled poly bags with manufacturing lot number and Date of Manufacture for traceability
Designed for higher temperature steam sterilization cycles (135 °C)* and compatible with ethylene oxide and hydrogen peroxide sterilization.
Certifications: Certificates of Conformance sent with every shipment.
Frequently Bought Together