Orbit™ Molecular Models Class Kit
Orbit™ Molecular Models Class Kit
Orbit™ molecular models class kit, Atoms: 9 mm Ø centers; Covalent bonds: 3.5 cm L
  470325-256
Supplier #:  2375
Orbit™ Molecular Models Class Kit
  470325-256
Orbit™ Molecular Models Class Kit
Orbit™ Molecular Models Class Kit
Orbit™ molecular models class kit, Atoms: 9 mm Ø centers; Covalent bonds: 3.5 cm L
106,75 $
Kit of 1
With these molecular model kits, students learn about the structure and bonding of simple organic and inorganic molecules through teacher-led activities or independent learning.
  • 12 packets of Orbit™ molecular model components to build models of small organic and inorganic molecules
  • Atom centres accept bonds at the proper angles and can be reused time and again
  • Teacher and student guides included with procedure and suggested study questions
  • For use individually or in small groups of students
Procedure explores the structure and bonding of molecules in relation to electron dot diagrams and structural formulas.
Ordering information: Activities can be extended with the Orbit Organic Chemistry Molecular Models Class Kit.
Delivery information: Each kit contains 12 sets of these Orbit™ molecular building system parts: 2 Carbon atoms; 2 Nitrogen atoms; 6 Chlorine atoms; 4 Oxygen atoms; 1 Sulfur atom; 8 Hydrogen atoms; 20 Covalent bonds; 4 Flexible covalent bonds; and teachers guide and reproducible student worksheet.
  • Désignation:
    Orbit™ molecular models class kit
  • Dimensions:
    Atoms: 9 mm Ø centers; Covalent bonds: 3.5 cm L