About this item
This antibody has been tested to bind to Ebola Sudan strain (SUDV) GP protein (Catalog: B78A28). It targets GP1/GP2 interface. It was also tested to not bind to Ebola Zaire Makona strain GP protein (Catalog: P54P89). It was tested to neutralize VSV-Pseudovirus_Ebola Sudan Strain GP with Luciferase Reporter (Catalog: U95M81).
Type: Primary
Antigen: Glycoprotein
Clonality: Monoclonal
Clone: RV12
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG1 Lambda
Reactivity: Human
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76729-536
- 76729-498
- Antigen symbol:
- Ebola
- Ebola
- Antigen name:
- Glycoprotein
- Glycoprotein
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- RV12
- RV12
- Reactivity:
- Human
- Human
- Host:
- Human
- Human
- Isotype:
- IgG1 lambda
- IgG1 lambda
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Neutralisation:
- Yes
- Yes
- Modification:
- Recombinant monoclonal antibody
- Recombinant monoclonal antibody
- Molecular weight:
- 150 kDa
- 150 kDa
- Purification:
- Purified by Protein A chromatography
- Purified by Protein A chromatography
- Concentration:
- 1 mg/ml by refractometry
- 1 mg/ml by refractometry
- Storage buffer:
- PBS, pH 7.2
- PBS, pH 7.2
- Storage temperature:
- Store at 4 °C, −20 °C
- Store at 4 °C, −20 °C
- Shipping temperature:
- Ice
- Ice