Anti-Sarilumab Mouse Monoclonal Antibody [clone: 20F5]
Catalogue # 76510-108
Fournisseur: Genscript
Spécifications
- Antibody type:Primary
- Antigen name:Sarilumab
- Antigen symbol:Sarilumab
- Clonality:Monoclonal
- Clone:20F5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Size:40 µg
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS, pH 7.4, containing 0.02% sodium azide
- Storage temperature:The lyophilized product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2 - 8 °C or for up to 12 months at −20 °C or below. Avoid repeated freezing and thawing cycles
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml
- Shipping temperature:Ice
- Immunogen:Sarilumab
- Purification:Protein A affinity column
- Cat. no.:76510-108
- Packaging:Vial
Spécifications
À propos de cet article
The product is specific for sarilumab.
- Anti idiotype antibodies
- High specificit
- High affinity
- High stability
GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.
.
Type: Primary
Antigen: Sarilumab
Clonality: Monoclonal
Clone: 20F5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: