Anti-Nivolumab Rabbit Monoclonal Antibody (Biotin) [clone: 40F11]
76510-002
- Antibody type:Primary
- Antigen name:Nivolumab
- Antigen symbol:Nivolumab
- Clonality:Monoclonal
- Clone:40F11
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Size:40 µg
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS, pH 7.4, containing 1% BSA and 0.02% sodium azide
- Storage temperature:The lyophilized product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2 - 8 °C or for up to 12 months at −20 °C or below. Avoid repeated freezing and thawing cycles
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml
- Shipping temperature:Ice
- Immunogen:Nivolumab
- Purification:Protein A affinity column
- Cat. no.:76510-002
- Packaging:Vial
The antibody is recommended as a detection antibody in a pharmacokinetic (PK) bridging assay with capture antibody GenScript, A01987-40, MonoRab anti-nivolumab antibody (33H4), mAb, rabbit.
- Anti idiotype antibodies
- High specificit
- High affinity
- High stability
GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.
.
Type: Primary
Antigen: Nivolumab
Clonality: Monoclonal
Clone: 40F11
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: