Anti-c-Myc Tag Rabbit Polyclonal Antibody
Catalog # 76197-198
Supplier: Genscript
Specifications
- Antibody type:Tag
- Antigen name:c-Myc Tag
- Antigen symbol:c-Myc Tag
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Western blot:Yes
- Size:40 µg
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS, pH 7.4 containing 0.02% sodium azide
- Storage temperature:The antibody is stable in lyophilized form if stored at −20 °C or below.The reconstituted antibody can be stored for 2 - 3 weeks at 2 - 8° C. For long term storage, aliquot and store at −20 °C or below. Avoid repeated freezing and thawing cycles
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml
- Shipping temperature:Ice
- Immunogen:c-Myc epitope tag peptide EQKLISEEDL conjugated to KLH
- Purification:Affinity chromatography
- Cat. no.:76197-198
- Packaging:Vial
Specifications
About this item
This antibody recognizes c-Myc tagged fusion proteins.
- Comprehensive pool
- against 24 tags
- Available in gram level bulk quantities
- Various conjugates for hot tags detection
GenScript offers more than 90 highly specific monoclonal and polyclonal antibodies against over 30 different tags. They are pre-validated for a variety of immunological applications, such as western blot analysis, immunoprecipitation, immunofluorescence, and affinity purification. Our epitope tag antibodies are manufactured under extensive quality control to ensure optimal sensitivity and consistent high performance.
Type: Tag
Antigen: c-Myc Tag
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: