Order Entry
Canada
ContactUsLinkComponent
Anti-NWSHPQFEK Tag Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase)) [clone: ]
Catalogue # 76198-728
Fournisseur:  Genscript
Anti-NWSHPQFEK Tag Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase)) [clone: ]
Catalogue # 76198-728
Fournisseur:  Genscript
Supplier Number:  A00875-40

Spécifications

  • Antibody type:
    Tag
  • Antigen name:
    NWSHPQFEK Tag
  • Antigen symbol:
    NWSHPQFEK Tag
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Size:
    40 µg
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Storage buffer:
    PBS, pH 7.4 containing 1% BSA and 0.01% thimerosal
  • Storage temperature:
    The antibody is stable in lyophilized form if stored at −20 °C or below.The reconstituted antibody can be stored for 2 - 3 weeks at 2 - 8 °C.For long term storage, aliquot and store at −20 °C or below.Avoid repeated freezing and thawing cycles
  • Concentration:
    Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Strep-tag II epitope peptide NWSHPQFEK conjugated to KLH.
  • Purification:
    Affinity chromatography
  • Cat. no.:
    76198-728
  • Packaging:
    Vial

Spécifications

À propos de cet article

This antibody recognizes strep-tag II tagged fusion proteins.

  • Comprehensive pool
  • against 24 tags
  • Available in gram level bulk quantities
  • Various conjugates for hot tags detection

GenScript offers more than 90 highly specific monoclonal and polyclonal antibodies against over 30 different tags. They are pre-validated for a variety of immunological applications, such as western blot analysis, immunoprecipitation, immunofluorescence, and affinity purification. Our epitope tag antibodies are manufactured under extensive quality control to ensure optimal sensitivity and consistent high performance.

Type: Tag
Antigen: NWSHPQFEK Tag
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: