Anti-Adalimumab Mouse Monoclonal Antibody [clone: 3C2] (Biotin)
76509-940
- Antibody type:Primary
- Antigen name:Adalimumab
- Antigen symbol:Adalimumab
- Clonality:Monoclonal
- Clone:3C2
- Conjugation:Biotin
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Size:40 µg
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS, pH 7.4, contains 1% BSA and 0.02% sodium azide
- Storage temperature:The lyophilized product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2 - 8 °C or for up to 12 months at −20 °C or below. Avoid freeze/thaw cycles
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml
- Shipping temperature:Ice
- Immunogen:Adalimumab
- Purification:Protein A affinity column
- Cat. no.:76509-940
- Packaging:Vial
The product is specific for Adalimumab. The antibody is recommended as a detection antibody in a pharmacokinetic (PK) bridging assay with capture antibody GenScript, A01954-40, Anti- Adalimumab Antibody (15C7), mAb, Mouse.
- Anti-Idiotype antibodies
- High specificit
- High affinity
- High stability
GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.
Type: Primary
Antigen: Adalimumab
Clonality: Monoclonal
Clone: 3C2
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: