Anti-Adalimumab Rabbit Polyclonal Antibody
76197-336
- Antibody type:Primary
- Antigen name:Adalimumab
- Antigen symbol:Adalimumab
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Size:40 µg
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS, pH 7.4, containing 0.02% sodium azide
- Storage temperature:The lyophilized product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2 - 8 °C or for up to 12 months at −20 °C or below. Avoid repeated freezing and thawing cycles
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml
- Shipping temperature:Ice
- Immunogen:Adalimumab
- Purification:Antigen affinity purification
- Cat. no.:76197-336
- Packaging:Vial
The product is specific for Adalimumab. This antibody serves as an excellent positive control for Adalimumab The product is specific for Adalimumab. This antibody serves as an excellent positive control for Adalimumab immunogenicity (ADA) assays.
- Anti-Idiotype antibodies
- High specificit
- High affinity
- High stability
GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.
Type: Primary
Antigen: Adalimumab
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: