Order Entry
Canada
ContactUsLinkComponent
Anti-Camelid VHH Rabbit Monoclonal Antibody [clone: 96A3F5] (iFluor 488)
Catalogue # 76197-320
Fournisseur:  Genscript
Anti-Camelid VHH Rabbit Monoclonal Antibody [clone: 96A3F5] (iFluor 488)
Catalogue # 76197-320
Fournisseur:  Genscript
Supplier Number:  A01862-200

Spécifications

  • Antibody type:
    Primary
  • Antigen name:
    Camelid VHH
  • Antigen symbol:
    Camelid VHH
  • Clonality:
    Monoclonal
  • Clone:
    96A3F5
  • Conjugation:
    iFluor® 488
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Camel,
    Llama,
    Alpaca
  • Western blot:
    Yes
  • Size:
    200 µl
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Storage buffer:
    PBS (pH 7.4), 10 mg/ml BSA, 0.02% sodium azide and 50% glycerol
  • Storage temperature:
    Store at −20 °C. This product is stable for 1 year upon receipt, when handled and stored as instructed
  • Concentration:
    0.5 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Llama VHH antibody
  • Purification:
    Protein A affinity column
  • Cat. no.:
    76197-320
  • Packaging:
    Vial

Spécifications

À propos de cet article

GenScript MonoRab™ Rabbit Anti-Camelid VHH Antibody [iFluor488], mAb is specific for Camelid VHH Antibody (Alpaca, Llama and Camel). The monoclonal antibody has no cross-reactivity with mouse, rat, rabbit, chicken, goat or human immunoglobulins.

  • Anti-VHH Antibodies
  • MonoRab™ technology guarantees high affinity: Kd ~10 to 11 M
  • No cross reactivity with other species
  • Specific to camelid IgG2 and IgG3
  • Recognize conformational epitopes of the VHH domain
  • Recognize both camelid and humanized VHHs

GenScript has developed a series of rabbit monoclonal (MonoRab) anti-VHH antibodies to facilitate your research of VHHs. Our anti-VHH antibodies recognize conformational epitopes of the VHH domain of camelid lgG2 and lgG3 subclasses. It is not necessary to add a tag to VHH for detection anymore.

Type: Primary
Antigen: Camelid VHH
Clonality: Monoclonal
Clone: 96A3F5
Conjugation: iFluor 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Alpaca;Llama;Camel