Hanna Instruments Spare pH Sensor for Hanna Testers
À propos de cet article
This spare pH sensor makes electrode replacement quick and easy. The spare pH sensor is designed as an easy-fit cartridge that locks into place, featuring two o-rings to maintain a watertight seal. With no pins to line up or that can break, this sensor is ideal for quick replacements.
- Easy-to-replace
- Extend the life of your Hanna pH tester with a spare pH sensor
- Keep your pH measurements simple and accurate
Replacing a pH sensor on one of Hanna's compatible testers is both quick and easy. The spare pH sensor is designed as an easy-fit cartridge that locks into place, featuring two o-rings to maintain a watertight seal. Replacement takes a few seconds and does not have any delicate pins that could break.
Ordering information: Replacement pH sensor for certain Hanna pH testers. Cannot use this for any pH meter. pH electrode storage solutions, pH calibration solutions, and pH electrode cleaning solutions sold seperately.
1 Options Available Below
Product Details & Documents
This spare pH sensor makes electrode replacement quick and easy. The spare pH sensor is designed as an easy-fit cartridge that locks into place, featuring two o-rings to maintain a watertight seal. With no pins to line up or that can break, this sensor is ideal for quick replacements.
- Easy-to-replace
- Extend the life of your Hanna pH tester with a spare pH sensor
- Keep your pH measurements simple and accurate
Replacing a pH sensor on one of Hanna's compatible testers is both quick and easy. The spare pH sensor is designed as an easy-fit cartridge that locks into place, featuring two o-rings to maintain a watertight seal. Replacement takes a few seconds and does not have any delicate pins that could break.
Ordering information: Replacement pH sensor for certain Hanna pH testers. Cannot use this for any pH meter. pH electrode storage solutions, pH calibration solutions, and pH electrode cleaning solutions sold seperately.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: HI73127Spécifications:New ProductNew Product
- Spare pH sensor for Hanna testersDiscontinued-Supplier Number: HI73127