Hanna Instruments Spare pH Sensor for Hanna Testers | Avantor
Hanna Instruments Spare pH Sensor for Hanna Testers
Supplier:   Hanna
Hanna Instruments Spare pH Sensor for Hanna Testers
Supplier:  Hanna
Certificates
Hanna Instruments Spare pH Sensor for Hanna Testers
Hanna Instruments Spare pH Sensor for Hanna Testers
1 Options Available Below
Product Details & Documents
This spare pH sensor makes electrode replacement quick and easy. The spare pH sensor is designed as an easy-fit cartridge that locks into place, featuring two o-rings to maintain a watertight seal. With no pins to line up or that can break, this sensor is ideal for quick replacements.
  • Easy-to-replace
  • Extend the life of your Hanna pH tester with a spare pH sensor
  • Keep your pH measurements simple and accurate
Replacing a pH sensor on one of Hanna's compatible testers is both quick and easy. The spare pH sensor is designed as an easy-fit cartridge that locks into place, featuring two o-rings to maintain a watertight seal. Replacement takes a few seconds and does not have any delicate pins that could break.
Ordering information: Replacement pH sensor for certain Hanna pH testers. Cannot use this for any pH meter. pH electrode storage solutions, pH calibration solutions, and pH electrode cleaning solutions sold seperately.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Spare pH sensor for Hanna testers
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $147.95
    $147.95
    Each
       $147.95
    Supplier Number: HI73127
    Spécifications:
    New Product
    New Product
  • Spare pH sensor for Hanna testers
    Discontinued
    Supplier Number: HI73127
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.