Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Hanna Instruments General Purpose Cleaning Solution (230 ml) | Avantor

Hanna Instruments General Purpose Cleaning Solution (230 ml)

Supplier:   Hanna
Hanna Instruments General Purpose Cleaning Solution (230 ml)
Supplier:  Hanna
Certificates
Hanna Instruments General Purpose Cleaning Solution (230 ml)
Hanna Instruments General Purpose Cleaning Solution (230 ml)

1 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

This general purpose cleaning solution can be used for a variety of applications. Electrodes can become dirty from use and will produce inaccurate results even as they read correctly in a pH buffer. Hanna's cleaning solutions eliminate impurities and residues that are left on electrode surfaces when immersed in samples during measurement or when stored incorrectly. Hanna suggests cleaning the bulb and junction of your electrode on a regular basis to ensure that the probe is always clean and to prevent any clogging of the junction.

  • Specially formulated for cleaning electrodes
  • Air tight seal to ensure quality

The most common cause for pH measurement inaccuracies is an unclean or improperly cleaned electrode. This is very important to note, because during calibration, the instrument assumes that the electrode is clean and that the standardization curve created during the calibration process will remain a valid reference until the next calibration. pH meters on the market today will allow an offset voltage of approximately ±60 mV. The deviation from 0 mV is not unusual but ideally should be no greater than ±30 mV. The calibration process compensates for the change in offset voltage. If the mV offset continues to deviate and the meter is calibrated with a dirty electrode then inaccurate readings will result.

Cleaning is a fast and effective routine that should be performed on a regular basis as a preventative measure against using a dirty electrode and to ensure that the junction is not clogged. By simply immersing the electrode in the HI7061M cleaning solution for 15 to 20 minutes will dissolve any mineral deposits or other coatings.

Product Family Options

Informations sur le produit

  • Désignation
    Availability
    Price
  • General Purpos Cleaning Solution (230 ml)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $22.88
    $22.88
    Each
       $22.88

    Supplier Number: HI7061M
    Spécifications:
    New Product
    New Product
  • General Purpos Cleaning Solution (230 ml)
    Discontinued
    Supplier Number: HI7061M

Certains produits peuvent apparaître comme restreints

Pour garantir un passage en caisse fluide et rapide,veuillez-vous connecter à votre compte.Certains articles peuvent apparaître comme restreints simplement parce que vous n'êtes pas connecté.

Si vous n'avez pas de compte, vous pouvez vous inscrire en utilisant notre formulaire d'inscription en ligne (https://www.avantorsciences.com/ca/en/login/register)

 

Si des restrictions apparaissent toujours après votre connexion, certains produits — comme les produits chimiques ou les dispositifs médicaux — nécessitent des étapes supplémentaires de vérification de compte pour pouvoir passer une commande. Certains articles peuvent également nécessiter une licence spécifique ou une documentation supplémentaire ; la documentation requise sera demandée pour ces articles avant l’expédition.

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.