Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Hanna Instruments Salinity Test Kit | Avantor

Hanna Instruments Salinity Test Kit

Supplier:   Hanna
Hanna Instruments Salinity Test Kit
Supplier:  Hanna
Certificates
Hanna Instruments Salinity Test Kit

1 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

This chemical test kit that measures salinity by mercuric nitrate titration. This is supplied complete with all of the reagents and equipment necessary to perform approximately 110 tests.

  • Pre-made reagents for ease of use
  • All reagents marked with expiration date and lot number for traceability
  • Manual titration performed with diphenylcarbazone indicator

This is a titration-based chemical test kit that measures salinity within the 0.0 to 40.0 g/kg range. The HI3835 is supplied with all of the necessary reagents and equipment to perform the analysis. The test kit contains enough reagents for perform approximately 110 tests. Salinity is defined as the total solids in water after all carbonates have been converted to oxides, all bromide and iodide have been replaced by chloride, and all organic matter has been oxidized. The salinity value is in g/kg or ppt (parts per thousand). The monitoring of salinity is essential for industrial waste and seawater, as different species of plants and animals thrive varying salinity levels.

Ordering information: Replacement reagents need to be purchased seperatly.

Caution: This product can expose you to chemicals including mercury and mercury compounds, which are known to the State of California to cause cancer. (Propisition 65)

Specifications for Product Family

Spécifications

Spécifications

  • Catalog No:
  • CA470006-748
  • 470006-748
  • Method:
  • Mercuric nitrate
  • Mercuric nitrate

Spécifications

Product Family Options

Informations sur le produit

  • Désignation
    Range
    No. of Tests
    Availability
    Price
  • Salinity test kit
    0 - 40 g/kg (ppt)
    110 average
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $102.75
    $102.75
    Each
       $102.75

    Supplier Number: HI3835
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    CA470006-748
    Method
    Mercuric nitrate
    New Product
  • Salinity test kit
    0 - 40 g/kg (ppt)
    110 average
    Discontinued
    Supplier Number: HI3835
    Spécifications:
    More
    Specifications
    Cat. No.
    470006-748
    Method
    Mercuric nitrate

Certains produits peuvent apparaître comme restreints

Pour garantir un passage en caisse fluide et rapide,veuillez-vous connecter à votre compte.Certains articles peuvent apparaître comme restreints simplement parce que vous n'êtes pas connecté.

Si vous n'avez pas de compte, vous pouvez vous inscrire en utilisant notre formulaire d'inscription en ligne (https://www.avantorsciences.com/ca/en/login/register)

 

Si des restrictions apparaissent toujours après votre connexion, certains produits — comme les produits chimiques ou les dispositifs médicaux — nécessitent des étapes supplémentaires de vérification de compte pour pouvoir passer une commande. Certains articles peuvent également nécessiter une licence spécifique ou une documentation supplémentaire ; la documentation requise sera demandée pour ces articles avant l’expédition.

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.