Celestron SkySync GPS Accessory
Skysnyc GPS accessory
Catalog # 470351-060
Supplier: Celestron International
Supplier #: 93969
Jump to:
Product Details & Documents
Add GPS to your Celestron computerized telescope and get time, date, and location downloads from orbiting Global Positioning Satellites.
- Upgrade your Celestron computerized telescope to GPS with SkySync
- Improve the speed and accuracy of alignments with 16-channel GPS
- Automatically updates the time and date
- Plugs into the AUX port on the base of the telescope
- Compatible for all non-GPS Celestron computerized telescopes
This 16-channel GPS module plugs into your telescope’s hand control or the auxiliary (AUX) port on your drive base and then mounts to a tripod leg to stay safely out of the way with included straps.
SkySync makes it easier to align your telescope while keeping your date and time updated.
Specifications
- Désignation:Skysnyc GPS accessory