Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Mouse Melatonin ELISA Kit | Avantor

Mouse Melatonin ELISA Kit

Mouse melatonin ELISA kit, 96 tests
Catalog # 76743-916
Supplier:   Antibodies.com
Fournisseur #:  A79543-96
Mouse Melatonin ELISA Kit
Catalog # 76743-916
Supplier:  Antibodies.com
CAS Number:  
Mouse Melatonin ELISA Kit
Mouse Melatonin ELISA Kit

Mouse melatonin ELISA kit, 96 tests

CAS Number:  
Price
$974.35
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

Mouse Melatonin ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of mouse Melatonin in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA kit
  • Detection Range: 7.813 to 500 pg/ml
  • Sensitivity: 4.688 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Mouse Melatonin ELISA kit (A79543) employs the competitive enzyme immunoassay technique for the quantitative measurement of mouse Melatonin in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Melatonin antigen. During the incubation, Melatonin present in the samples or standards competes with the fixed amount of immobilized Melatonin for binding sites on the Biotinylated Anti-Melatonin Antibody. The more Melatonin present in a sample or standard, the less Biotinylated Anti-Melatonin Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Melatonin Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Melatonin present in each sample or standard. The concentration of Melatonin can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Spécifications

Spécifications

  • Désignation:
    Mouse melatonin ELISA kit
  • Taille:
    96 tests
  • Primary antibody reactivity:
    Mouse
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Sample volume:
    50 μl
  • Detection range:
    7.813 - 500 pg/ml
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Assay Type:
    Competitive
  • Time to Results:
    4 h 30 min
  • Cross reactivity:
    Mouse Melatonin ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse Melatonin. No significant cross-reactivity or interference between Melatonin and analogues was observed.
  • Assay Principle:
    Quantitative
  • Cat. No.:
    76743-916
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    4.688 pg/ml
  • Format:
    Pre-coated
  • Target protein:
    Melatonin
  • Regulatory status:
    RUO

Spécifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.