Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Rat Thyroxine/T4 ELISA Kit | Avantor

Rat Thyroxine/T4 ELISA Kit

Rat thyroxine/T4 ELISA kit, 96 tests
Catalog # 76737-672
Supplier:   Antibodies.com
Fournisseur #:  A247022-96
Rat Thyroxine/T4 ELISA Kit
Catalog # 76737-672
Supplier:  Antibodies.com
CAS Number:  
Rat Thyroxine/T4 ELISA Kit
Rat Thyroxine/T4 ELISA Kit

Rat thyroxine/T4 ELISA kit, 96 tests

CAS Number:  
Price
$1,071.04
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

Rat Thyroxine/T4 ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of rat Thyroxine/T4 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Rat Thyroxine/T4 ELISA kit (A247022) employs the competitive enzyme immunoassay technique for the quantitative measurement of rat Thyroxine/T4 in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Thyroxine/T4 antigen. During the incubation, Thyroxine/T4 present in the samples or standards competes with the fixed amount of immobilized Thyroxine/T4 for binding sites on the Biotinylated Anti-Thyroxine/T4 Antibody. The more Thyroxine/T4 present in a sample or standard, the less Biotinylated Anti-Thyroxine/T4 Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Thyroxine/T4 Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Thyroxine/T4 present in each sample or standard. The concentration of Thyroxine/T4 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Spécifications

Spécifications

  • Désignation:
    Rat thyroxine/T4 ELISA kit
  • Taille:
    96 tests
  • Primary antibody reactivity:
    Rat
  • Sample type:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Sample volume:
    50 μl
  • Detection range:
    0.703 - 45 ng/ml
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Assay Type:
    Competitive
  • Time to Results:
    4 h 30 min
  • Cross reactivity:
    Rat Thyroxine/T4 ELISA Kit exhibits high specificity and excellent specificity for the detection of rat Thyroxine/T4. No significant cross-reactivity or interference between Thyroxine/T4 and analogues was observed.
  • Cat. No.:
    76737-672
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    0.422 ng/ml
  • Format:
    Pre-coated
  • Target protein:
    Thyroxine/T4
  • Regulatory status:
    RUO

Spécifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.