Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Progesterone ELISA Kit

Progesterone ELISA kit, 96 tests
Catalog # 76731-944
Supplier:   Antibodies.com
 :  A75712-96
Progesterone ELISA Kit
Catalog # 76731-944
Supplier:  Antibodies.com
CAS Number:  
Progesterone ELISA Kit
Progesterone ELISA Kit

Progesterone ELISA kit, 96 tests

CAS Number:  
Price
$974.35
Log in pour la disponibilité
Each
 
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

Progesterone ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of Progesterone in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 0.313 to 20 ng/ml
  • Sensitivity: 0.188 ng/ml
  • Sample type: Serum. plasma. tissue homogenates. and other biological fluids
  • Assay precision: Intra assay: CV <8%. Inter assay: CV <10%

Progesterone ELISA kit (A75712) employs the competitive enzyme immunoassay technique for the quantitative measurement of universal Progesterone in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Progesterone antigen. During the incubation, Progesterone present in the samples or standards competes with the fixed amount of immobilized Progesterone for binding sites on the Biotinylated Anti-Progesterone Antibody. The more Progesterone present in a sample or standard, the less Biotinylated Anti-Progesterone Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Progesterone Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Progesterone present in each sample or standard. The concentration of Progesterone can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Spécifications

Spécifications

  • Désignation:
    Progesterone ELISA kit
  • Taille:
    96 tests
  • Primary antibody reactivity:
    Other
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Sample volume:
    50 μl
  • Detection range:
    0.313 - 20 ng/ml
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Assay Type:
    Competitive
  • Time to Results:
    4 h 30 min
  • Cross reactivity:
    Progesterone ELISA kit exhibits high specificity and excellent specificity for the detection of universal Progesterone. No significant cross-reactivity or interference between Progesterone and analogues was observed.
  • Assay Principle:
    Quantitative
  • Cat. No.:
    76731-944
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    0.188 ng/ml
  • Format:
    Pre-coated
  • Target protein:
    Progesterone
  • Regulatory status:
    RUO

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.