Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Mouse Cortisol ELISA Kit | Avantor

Mouse Cortisol ELISA Kit

Mouse COR ELISA kit, 96 tests
Catalog # 76740-558
Supplier:   Antibodies.com
Fournisseur #:  A87503-96
Mouse Cortisol ELISA Kit
Catalog # 76740-558
Supplier:  Antibodies.com
CAS Number:  
Mouse Cortisol ELISA Kit
Mouse Cortisol ELISA Kit

Mouse COR ELISA kit, 96 tests

CAS Number:  
Price
1 071,04 $
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

Mouse Cortisol ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of mouse Cortisol in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA Kit
  • Detection Range: 0.391 to 25 ng/ml
  • Sensitivity: 0.234 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Mouse Cortisol ELISA Kit (A87503) employs the competitive enzyme immunoassay technique for the quantitative measurement of mouse Cortisol in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Cortisol antigen. During the incubation, Cortisol present in the samples or standards competes with the fixed amount of immobilized Cortisol for binding sites on the Biotinylated Anti-Cortisol Antibody. The more Cortisol present in a sample or standard, the less Biotinylated Anti-Cortisol Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Cortisol Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Cortisol present in each sample or standard. The concentration of Cortisol can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Spécifications

Spécifications

  • Désignation:
    Mouse COR ELISA kit
  • Taille:
    96 tests
  • Primary antibody reactivity:
    Mouse
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Sample volume:
    50 μl
  • Detection range:
    0.391 - 25 ng/ml
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Assay Type:
    Competitive
  • Time to Results:
    4 h 30 min
  • Cross reactivity:
    Mouse Cortisol ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse Cortisol. No significant cross-reactivity or interference between Cortisol and analogues was observed.
  • Assay Principle:
    Quantitative
  • Cat. No.:
    76740-558
  • Detection method:
    Colorimetric
  • Sensitivity:
    0.234 ng/ml
  • Format:
    Pre-coated
  • Target protein:
    COR
  • Regulatory status:
    RUO

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.