Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Estrone ELISA Kit | Avantor

Estrone ELISA Kit

Estrone ELISA kit, 96 tests
Catalog # 76731-538
Supplier:   Antibodies.com
Fournisseur #:  A73992-96
Estrone ELISA Kit
Catalog # 76731-538
Supplier:  Antibodies.com
CAS Number:  
Estrone ELISA Kit
Estrone ELISA Kit

Estrone ELISA kit, 96 tests

CAS Number:  
Price
$974.35
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

Estrone ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of Estrone in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA Kit
  • Detection Range: 0.156 to 10 pmol/ml
  • Sensitivity: 0.094 pmol/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Estrone ELISA Kit (A73992) employs the competitive enzyme immunoassay technique for the quantitative measurement of universal Estrone in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Estrone antigen. During the incubation, Estrone present in the samples or standards competes with the fixed amount of immobilized Estrone for binding sites on the Biotinylated Anti-Estrone Antibody. The more Estrone present in a sample or standard, the less Biotinylated Anti-Estrone Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Estrone Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Estrone present in each sample or standard. The concentration of Estrone can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Spécifications

Spécifications

  • Désignation:
    Estrone ELISA kit
  • Taille:
    96 tests
  • Primary antibody reactivity:
    Other
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Sample volume:
    50 μl
  • Detection range:
    0.156 - 10 pmol/ml
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Assay Type:
    Competitive
  • Time to Results:
    4 h 30 min
  • Cross reactivity:
    Estrone ELISA Kit exhibits high specificity and excellent specificity for the detection of universal Estrone. No significant cross-reactivity or interference between Estrone and analogues was observed.
  • Assay Principle:
    Quantitative
  • Cat. No.:
    76731-538
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    0.094 pmol/ml
  • Format:
    Pre-coated
  • Target protein:
    Estrone
  • Regulatory status:
    RUO

Spécifications