Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Human CD2 ELISA Kit | Avantor

Human CD2 ELISA Kit

Human CD2 ELISA kit, 96 tests
Catalog # 76743-780
Supplier:   Antibodies.com
Fournisseur #:  A79499-96
Human CD2 ELISA Kit
Catalog # 76743-780
Supplier:  Antibodies.com
CAS Number:  
Human CD2 ELISA Kit
Human CD2 ELISA Kit

Human CD2 ELISA kit, 96 tests

CAS Number:  
Price
$1,071.04
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

Human CD2 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human CD2 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-To-Use ELISA Kit
  • Detection Range: 0.313 - 20 ng/ml
  • Sensitivity: 0.188 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Human CD2 ELISA kit (A79499) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human CD2 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for CD2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the CD2 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-CD2 Antibody, which binds the captured CD2 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of CD2 captured in each well. The concentration of CD2 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Spécifications

Spécifications

  • Désignation:
    Human CD2 ELISA kit
  • Taille:
    96 tests
  • Primary antibody reactivity:
    Human
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Sample volume:
    100 μl
  • Detection range:
    0.313 - 20 ng/ml
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    4 h 30 min
  • Cross reactivity:
    Human CD2 ELISA kit exhibits high specificity and excellent specificity for the detection of human CD2. No significant cross-reactivity or interference between CD2 and analogues was observed.
  • Assay Principle:
    Quantitative
  • Cat. No.:
    76743-780
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    0.188 ng/ml
  • Format:
    Pre-coated
  • Target protein:
    CD2
  • Regulatory status:
    RUO

Spécifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.