5-Hydroxytryptamine ELISA Kit
5-Hydroxytryptamine ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of 5-Hydroxytryptamine in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA Kit
- Detection Range: 1.563 to 100 ng/ml
- Sensitivity: 0.938 ng/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
5-Hydroxytryptamine ELISA Kit (A74022) employs the competitive enzyme immunoassay technique for the quantitative measurement of universal 5-Hydroxytryptamine in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with 5-Hydroxytryptamine antigen. During the incubation, 5-Hydroxytryptamine present in the samples or standards competes with the fixed amount of immobilized 5-Hydroxytryptamine for binding sites on the Biotinylated Anti-5-Hydroxytryptamine Antibody. The more 5-Hydroxytryptamine present in a sample or standard, the less Biotinylated Anti-5-Hydroxytryptamine Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-5-Hydroxytryptamine Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of 5-Hydroxytryptamine present in each sample or standard. The concentration of 5-Hydroxytryptamine can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.
: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
- Désignation:5-Hydroxytryptamine ELISA kit
- Taille:96 tests
- Primary antibody reactivity:Other
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Sample volume:50 μl
- Detection range:1.563 - 100 ng/ml
- Storage temperature:4 °C
- Shelf life:Store for 6 months at 4 °C
- Assay Type:Competitive
- Time to Results:4 h 30 min
- Cross reactivity:5-Hydroxytryptamine ELISA Kit exhibits high specificity and excellent specificity for the detection of universal 5-Hydroxytryptamine. No significant cross-reactivity or interference between 5-Hydroxytryptamine and analogues was observed.
- Assay Principle:Quantitative
- Cat. No.:76731-450
- Assay duration:Multiple steps
- Detection method:Colorimetric
- Sensitivity:0.938 ng/ml
- Format:Pre-coated
- Target protein:5-Hydroxytryptamine
- Regulatory status:RUO
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



