Eisco® Aneroid Teaching Barometer | Avantor
Eisco® Aneroid Teaching Barometer
Aneroid teaching barometer
  470344-804
 :   Wards
Fournisseur #:  PH0248
Eisco® Aneroid Teaching Barometer
  470344-804
 :  Wards
Eisco® Aneroid Teaching Barometer
Eisco® Aneroid Teaching Barometer
Aneroid teaching barometer
$64.60
Each
Accurately measures barometric pressure and is ideal for classroom use.
  • 13 cm dial numbered in inches and millibar
  • Exposed aneroid shows change in pressure
  • From 28 to 31" of mercury (960 to 1060 millibar)
The EISCO aneroid teaching barometer with pump is a high quality teaching barometer to demonstrate operation. The exposed aneroid shows the change in barometric pressure when you pump the attached bulb.
  • Désignation:
    Aneroid teaching barometer