Eisco® High Resolution Dynamometer | Avantor
Eisco® High Resolution Dynamometer
High resolution dynamometer, 100 g/1 N
Catalog # 470344-060
Supplier:   Wards
Fournisseur #:  PH0036AAL
Eisco® High Resolution Dynamometer
Catalog # 470344-060
Supplier:  Wards
Eisco® High Resolution Dynamometer
Eisco® High Resolution Dynamometer
High resolution dynamometer, 100 g/1 N
Price
$20.30
Log in pour la disponibilité
Each
Product Details & Documents
High-resolution dynamometer with a capacity of 100 g/1N. Can measure both grams and Newtons. Ideal for laboratory experiments and measuring small forces.
  • Aluminum
  • Capacity: 0 to 100 g and 0 to 1N
  • 8.75" length (with no weight attached)
  • Silver metallic color
  • High resolution
This 100 g/1N high resolution dynamometer made of aluminum is a great tool for accurately assessing the weight of an object in both grams and Newtons. It has easy to read markings and is very user-friendly.
Spécifications
  • Capacité:
    100 g/1 N
  • Désignation:
    High resolution dynamometer