Eisco® High Resolution, Spring Balances with Dual Scale
Supplier: Wards
Certificates
About this item
High-resolution spring balance with transparent dual scale in Newtons and grams. Zero calibration capability and acrylic body for durability.
- Premium quality spring balances
- Acrylic body with dual transparent scale - measures in Newtons and grams
- User friendly zero-point calibration capability
- Ideal for physics demonstrations and experiments
- Spring balance scale measures 9" tall in total, 1.4" wide and 0.85" long
User friendly zero-point calibration capability- simply pull up or push down the tab at the top of the scale to adjust. Features a loop at the top for hanging and a hook at the base to hold items being measured. Ideal for physics classrooms, demonstrations and experiments. Height of the scale body is 5.4".
Frequently Bought Together