TableWorx Performance Work Tables
TableWorx Performance Work Tables
Supplier:   Metro
TableWorx Performance Work Tables
Supplier:  Metro
Certificates
TableWorx Performance Work Tables
TableWorx Performance Work Tables
29 Options Available Below
Product Details & Documents
Metro TableWorx is a truly modular platform of performance work tables designed to allow you to customize your perfect solution.
  • Innumerable options and a truly modular platform allow you to customize a perfect solution
  • Overhead riser solution can be added at anytime converting it into a productivity workcenter
  • Work surface alloy: type 304 or 316 stainless steel
  • Work surface load capacity of 650 lbs.
  • Height adjustability
All TableWorx Work Tables feature a 16-gauge, Type 304 or 316 stainless steel work surface reinforced with 16-gauge hat channels. The sound deadened work surface features 90-degree turned down edges on all four sides.
Out-of-the box, TableWorx can be assembled to any one of six working surface heights: 34" (864 mm), 35" (889 mm), 36" (914 mm), 37" (940 mm), 38" (965 mm), and 39" (991 mm). Stationary work table legs include stainless steel leveling bullet feet with 1.125" (29 mm) range of adjustability. Mobile-ready tables are delivered without casters.
Elevate your efficiency to new levels with the addition of TableWorx productivity riser. Risers can be easily added to any TableWorx work table. A full array of overshelves, wire grids, and accessories can be added to maximize every inch of space, making you more comfortable and productive than ever before. Risers can be mounted at the rear or center of the work table.
Certifications: NSF Certified.
Product Family Options
316 Stainless series
Product Information
  • Désignation
    Worksurface Material
    Base/Leg Material
    W×D
    Shipping Weight
    Availability
    Price
  • Mobile-ready worktable with backsplash (casters not included) with I-frame
    Stainless steel 316
    Stainless steel 304
    60×30"
    120.1 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $6,454.85
    $6,454.85
    Each
       $6,454.85
    Minimum order quantity is 1
    Supplier Number: TWM3060SI316BS
  • Mobile-ready worktable (casters not included) with I-frame
    Stainless steel 316
    Stainless steel 304
    60×36"
    122.8 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $6,357.01
    $6,357.01
    Each
       $6,357.01
    Minimum order quantity is 1
    Supplier Number: TWM3660SI316S
  • Stationary worktable with I-frame
    Stainless steel 316
    Stainless steel 304
    36×30"
    84.8 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $4,942.37
    $4,942.37
    Each
       $4,942.37
    Minimum order quantity is 1
    Supplier Number: TWS3036SI316S
  • Stationary worktable with 3-sided frame
    Stainless steel 316
    Stainless steel 304
    36×36"
    88.4 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $5,206.55
    $5,206.55
    Each
       $5,206.55
    Minimum order quantity is 1
    Supplier Number: TWS3636SU316S
  • Stationary worktable with backsplash with 3-sided frame
    Stainless steel 316
    Stainless steel 304
    72×30"
    142.3 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $7,041.81
    $7,041.81
    Each
       $7,041.81
    Minimum order quantity is 1
    Supplier Number: TWS3072SU316BS
  • Stationary worktable with backsplash with undershelf
    Stainless steel 316
    Stainless steel 304
    72×30"
    161.6 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $7,540.76
    $7,540.76
    Each
       $7,540.76
    Minimum order quantity is 1
    Supplier Number: TWS3072FS316BS
  • Stationary worktable with undershelf
    Stainless steel 316
    Stainless steel 304
    96×24"
    168.9 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $7,108.34
    $7,108.34
    Each
       $7,108.34
    Minimum order quantity is 1
    Supplier Number: TWS2496FS316S
All Stainless series
Product Information
  • Désignation
    Worksurface Material
    Base/Leg Material
    W×D
    Shipping Weight
    Availability
    Price
  • Mobile-ready worktable with backsplash (casters not included) with 3-sided frame
    Stainless steel 304
    Stainless steel 304
    60×36"
    131.6 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $5,979.37
    $5,979.37
    Each
       $5,979.37
    Minimum order quantity is 1
    Supplier Number: TWM3660SU304BS
  • Mobile-ready worktable with backsplash (casters not included) with 3-sided frame
    Stainless steel 304
    Stainless steel 304
    72×30"
    136.9 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $6,053.75
    $6,053.75
    Each
       $6,053.75
    Minimum order quantity is 1
    Supplier Number: TWM3072SU304BS
  • Mobile-ready worktable (casters not included) with I-frame
    Stainless steel 304
    Stainless steel 304
    72×30"
    127 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $5,466.76
    $5,466.76
    Each
       $5,466.76
    Minimum order quantity is 1
    Supplier Number: TWM3072SI304S
  • Stationary worktable with 3-sided frame
    Stainless steel 304
    Stainless steel 304
    24×24"
    63.5 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $3,744.95
    $3,744.95
    Each
       $3,744.95
    Minimum order quantity is 1
    Supplier Number: TWS2424SU304S
  • Stationary worktable with 3-sided frame
    Stainless steel 304
    Stainless steel 304
    60×24"
    104.8 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $4,690.01
    $4,690.01
    Each
       $4,690.01
    Minimum order quantity is 1
    Supplier Number: TWS2460SU304S
  • Stationary worktable with backsplash with I-frame
    Stainless steel 304
    Stainless steel 304
    36×36"
    99.2 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $4,926.74
    $4,926.74
    Each
       $4,926.74
    Minimum order quantity is 1
    Supplier Number: TWS3636SI304BS
Standard series
Product Information
  • Désignation
    Worksurface Material
    Base/Leg Material
    W×D
    Shipping Weight
    Availability
    Price
  • Mobile-ready worktable (casters not included) with I-frame
    Stainless steel 304
    Stainless steel 304 frame / epoxy coated legs with reinforced nylon polymer leg mounts
    36×30"
    73.7 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $2,948.63
    $2,948.63
    Each
       $2,948.63
    Minimum order quantity is 1
    Supplier Number: TWM3036SI304K
  • Mobile-ready worktable (casters not included) with I-frame
    Stainless steel 304
    Stainless steel 304 frame / epoxy coated legs with reinforced nylon polymer leg mounts
    72×30"
    121.3 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $3,975.83
    $3,975.83
    Each
       $3,975.83
    Minimum order quantity is 1
    Supplier Number: TWM3072SI304K
  • Mobile-ready worktable (casters not included) with 3-sided frame
    Stainless steel 304
    Stainless steel 304 frame / epoxy coated legs with reinforced nylon polymer leg mounts
    72×24"
    107.6 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $3,563.00
    $3,563.00
    Each
       $3,563.00
    Minimum order quantity is 1
    Supplier Number: TWM2472SU304K
  • Mobile-ready worktable (casters not included) with undershelf
    Stainless steel 304
    Stainless steel 304 frame / epoxy coated legs with reinforced nylon polymer leg mounts
    72×30"
    140.2 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $4,408.25
    $4,408.25
    Each
       $4,408.25
    Minimum order quantity is 1
    Supplier Number: TWM3072FS304K
  • Mobile-ready worktable with backsplash (casters not included) with 3-sided frame
    Stainless steel 304
    Stainless steel 304 frame / epoxy coated legs with reinforced nylon polymer leg mounts
    30×24"
    64.2 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $2,776.41
    $2,776.41
    Each
       $2,776.41
    Minimum order quantity is 1
    Supplier Number: TWM2430SU304BK
  • Mobile-ready worktable with backsplash (casters not included) with 3-sided frame
    Stainless steel 304
    Stainless steel 304 frame / epoxy coated legs with reinforced nylon polymer leg mounts
    36×36"
    87.8 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $3,490.60
    $3,490.60
    Each
       $3,490.60
    Minimum order quantity is 1
    Supplier Number: TWM3636SU304BK
  • Mobile-ready worktable with backsplash (casters not included) with 3-sided frame
    Stainless steel 304
    Stainless steel 304 frame / epoxy coated legs with reinforced nylon polymer leg mounts
    48×24"
    87.2 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $3,308.66
    $3,308.66
    Each
       $3,308.66
    Minimum order quantity is 1
    Supplier Number: TWM2448SU304BK
  • Mobile-ready worktable with backsplash (casters not included) with undershelf
    Stainless steel 304
    Stainless steel 304 frame / epoxy coated legs with reinforced nylon polymer leg mounts
    24×24"
    60.1 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $2,788.21
    $2,788.21
    Each
       $2,788.21
    Minimum order quantity is 1
    Supplier Number: TWM2424FS304BK
  • Mobile-ready worktable with backsplash (casters not included) with undershelf
    Stainless steel 304
    Stainless steel 304 frame / epoxy coated legs with reinforced nylon polymer leg mounts
    96×30"
    192.6 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $5,940.22
    $5,940.22
    Each
       $5,940.22
    Minimum order quantity is 1
    Supplier Number: TWM3096FS304BK
  • Stationary worktable with 3-sided frame
    Stainless steel 304
    Stainless steel 304 frame / epoxy coated legs with reinforced nylon polymer leg mounts
    48×24"
    85.3 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $3,009.26
    $3,009.26
    Each
       $3,009.26
    Minimum order quantity is 1
    Supplier Number: TWS2448SU304K
  • Stationary worktable with 3-sided frame
    Stainless steel 304
    Stainless steel 304 frame / epoxy coated legs with reinforced nylon polymer leg mounts
    48×30"
    94.9 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $3,294.91
    $3,294.91
    Each
       $3,294.91
    Minimum order quantity is 1
    Supplier Number: TWS3048SU304K
  • Stationary worktable with backsplash with 3-sided frame
    Stainless steel 304
    Stainless steel 304 frame / epoxy coated legs with reinforced nylon polymer leg mounts
    96×24"
    153.6 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $4,758.43
    $4,758.43
    Each
       $4,758.43
    Minimum order quantity is 1
    Supplier Number: TWS2496SU304BK
  • Stationary worktable with backsplash with I-frame
    Stainless steel 304
    Stainless steel 304 frame / epoxy coated legs with reinforced nylon polymer leg mounts
    96×24"
    154.1 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $4,803.40
    $4,803.40
    Each
       $4,803.40
    Minimum order quantity is 1
    Supplier Number: TWS2496SI304BK
  • Stationary worktable with backsplash with undershelf
    Stainless steel 304
    Stainless steel 304 frame / epoxy coated legs with reinforced nylon polymer leg mounts
    96×24"
    176.1 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $5,151.74
    $5,151.74
    Each
       $5,151.74
    Minimum order quantity is 1
    Supplier Number: TWS2496FS304BK
  • Stationary worktable with undershelf
    Stainless steel 304
    Stainless steel 304 frame / epoxy coated legs with reinforced nylon polymer leg mounts
    48×36"
    119 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $3,846.66
    $3,846.66
    Each
       $3,846.66
    Minimum order quantity is 1
    Supplier Number: TWS3648FS304K
  • Stationary worktable with undershelf
    Stainless steel 304
    Stainless steel 304 frame / epoxy coated legs with reinforced nylon polymer leg mounts
    96×24"
    163.3 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $4,576.50
    $4,576.50
    Each
       $4,576.50
    Minimum order quantity is 1
    Supplier Number: TWS2496FS304K
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Productivity riser with 2 center cantilevered, stainless steel type 304, 60" long
    Productivity riser with 2 center cantilevered, stainless steel type 304, 60" long
    76779-740
    Log in for availability
    Each - $2,750.00
Product Information
  • Catalog #
    For Use With
    Load Capacity
    Availability
    Price
  • Polymer/stainless total-guard caster (Total lock stem, thermopastic elastomer, gray, polymer/reinforced nylon rig; thermoplastic elastomer flat tread; polypropylene hub)
    Polymer/stainless total-guard caster (Total lock stem, thermopastic elastomer, gray, polymer/reinforced nylon rig; thermoplastic elastomer flat tread; polypropylene hub)
    76545-628
    Wire shelving
    250 lbs.
    Log in for availability
    Each - $265.00
Product Information
  • Catalog #
    Availability
    Price
  • Productivity riser with 2 rear cantilevered, stainless steel type 304, 60" long
    Productivity riser with 2 rear cantilevered, stainless steel type 304, 60" long
    76779-744
    Log in for availability
    Each - $2,950.00