Klercide™ Cleanroom Quat Metered Dose Concentrate Sterile
Catalog # CA76717-442
Supplier: ECOLAB INC CA
Specifications
- Description:Klercide™ Cleanroom quat metered dose concentrate sterile, double-bagged
- Size:4×1 gal.
- Cat. no.:CA76717-442
- Packaging:Bottle, capped
- Sterility:Sterile
- Form:Metered dose concentrate
Specifications
About this item
Klercide™ Cleanroom Quat Metered Dose Concentrate Sterile is a blend of quaternary ammonium compound blended with Water for Injection. Concentrated non-food contact sanitizer, disinfectant, virucide, and fungicide for hard, non-porous surfaces for use in cleanrooms and controlled areas.
- Formulated to leave minimal visible residues when used as a non-food contact sanitizer on hard, non-porous surfaces
- Is easy and economical to use
- Is 0.2 micron filtered, filled, and double bagged in a cleanroom environment
- Compatible with most surfaces found in a cleanroom
- Compatible with other Ecolab biocides for rotational use
Each 1-gallon container makes 50 gallons of diluted sanitizer (76 ml of concentrate makes 1 gallon of ready-to-use sanitizer). Flexible use instructions allow for product use as a sanitizer, disinfectant, or fungicidal application.
Health Canada DIN registered Sanitizer, Disinfectant, and Fungicide.
Certifications: DIN: 02497964