Klercide™ Cleanroom Quat Metered Dose Concentrate Non-Sterile
CA76717-454
- Description:Klercide™ Cleanroom quat metered dose concentrate, single-bagged
- Size:4×1 gal.
- Cat. no.:CA76717-454
- Packaging:Bottle, capped
- Sterility:Non sterile
- Form:Metered dose concentrate
Cleanroom Quat Metered Dose Concentrate Non-Sterile is a blend of quaternary ammonium compound and deionized water. Concentrated non-food contact sanitizer, disinfectant, and fungicide for hard, non-porous surfaces for use in cleanrooms and controlled areas.
- Formulated to leave minimal visible residues when used as a non-food contact sanitizer on hard, non-porous surfaces
- Is easy and economical to use
- Is 0.2 micron filtered, filled and double bagged in a cleanroom environment
- Non-corrosive for use in most applications
- Compatible with other Ecolab biocides for rotational use
Each 1-gallon container makes 50 gallons of diluted sanitizer (76 ml of concentrate makes 1 gallon of ready-to-use sanitizer). Flexible use instructions allow for product use as a sanitizer, disinfectant, or fungicidal application.
Health Canada DIN registered sanitizer, disinfectant, and fungicide.
: DIN: 02497964