CENCO® AP Physics Lab 1.11: Exploring a System in Rotation | Avantor
CENCO® AP Physics Lab 1.11: Exploring a System in Rotation
CENCO AP Physics Lab 1.11: Exploring a System in Rotation, 1
Catalog # 470348-766
Supplier:   Avantor
CENCO® AP Physics Lab 1.11: Exploring a System in Rotation
Catalog # 470348-766
Supplier:  Avantor
CENCO® AP Physics Lab 1.11: Exploring a System in Rotation
CENCO® AP Physics Lab 1.11: Exploring a System in Rotation
CENCO AP Physics Lab 1.11: Exploring a System in Rotation, 1
Price
$434.35
Log in pour la disponibilité
Kit of 1
Product Details & Documents
Optimize the distribution of weights on a wheel, then determine g using a rotational apparatus.
  • Investigation is aligned with the revised AP Physics standards from the College Board
  • Each kit is designed for a lab group of 2 to 4 students
  • Students consider meaningful, real-world objects, phenomena, and scenarios
  • Students compare classic experiments, drawing connections to abstract facts and equations
Introducing the new CENCO AP Physics lab series!
Ideal for AP Physics 1 and 2, AP Physics C, IB DP Physics, and advanced/honors physics courses, each of the 35 experiments is aligned with the revised College Board AP curriculum. These innovative labs go way beyond traditional exercises, turning real-world scenarios into true guided-inquiry investigations. Students are given a goal, but the investigation is driven by their ideas. As they draw conclusions, students report their results using the Claim-Evidence-Reasoning model. The teacher guides and student manuals are authored and reviewed by AP Physics teachers with decades of combined experience, so you can trust the setups to be successful in your classroom. All labs come complete with instructions and materials for one lab group of 2 to 4 students. For your convenience, labs are packaged to allow you to purchase only what is required for each experiment.
Moment of inertia is typically demonstrated using a big visual device or using a discrepant event rolling cylinders down a ramp. In this experiment, however, students must experiment with different distributions of mass on a rolling system and then intentionally design a configuration to minimize its acceleration. In part 2 of this investigation, students use a rotation apparatus to control a falling mass, thus discovering yet another way do determine the acceleration due to gravity: g.
Ordering information: Required but not included: Centigram balance, inclined plane, meter stick, stopwatch/smartphone.
Delivery information: Kit includes rotation apparatus with adjustable mass, CDs with rubber grommets, C clamp, modeling clay, wood dowel.
Spécifications
  • Désignation:
    CENCO AP Physics Lab 1.11: Exploring a System in Rotation
  • No. of Lab Groups:
    1