Order Entry
Canada
ContactUsLinkComponent
 
Anti-Lysyl tRNA synthetase Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # 76117-086
Supplier:  Bioss
Anti-Lysyl tRNA synthetase Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # 76117-086
Supplier:  Bioss
Supplier Number:  BS-6127R-A750
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Lysyl tRNA synthetase
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
  • Western blot:
    Yes
  • Size:
    100 µl
  • Environmentally Preferable:
  • Epitope:
    301-400/482
  • Form:
    Liquid
  • Gene ID:
    6850
  • Antigen synonyms:
    KRS|Lysine--tRNA ligase|LysRS|SYK_HUMAN|Lysyl-tRNA synthetase|KARS2|Lysine tRNA ligase|KARS|KARS 2|KIAA0070
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol.
  • Molecular weight:
    54 kDa
  • Storage temperature:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human KARS2/Lysyl tRNA syntase
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Cat. no.:
    76117-086
  • Packaging:
    Vial

Specifications

About this item

Catalyzes the specific attachment of an amino acid to its cognate tRNA in a 2 step reaction: the amino acid (AA) is first activated by ATP to form AA-AMP and then transferred to the acceptor end of the tRNA. When secreted, acts as a signaling molecule that induces immune response through the activation of monocyte/macrophages. Catalyzes the synthesis of diadenosine oligophosphate (Ap4A), a signaling molecule involved in the activation of MITF transcriptional activity. Interacts with HIV-1 virus GAG protein, facilitating the selective packaging of tRNA(3)(Lys), the primer for reverse transcription initiation.

Type: Primary
Antigen: Lysyl tRNA synthetase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 301-400/482
Host: Rabbit
Isotype: IgG
Reactivity: