ShuttlePouch, 95 kPa
ShuttlePouch, 95 kPa
ShuttlePouch, 95 kPa
ShuttlePouch, 95 kPa
ShuttlePouch, 95 kPa
Therapak’s 95 kPa transport pouches and bags are ideal to use as a secondary leakproof packaging in accordance with the Department of Transportation (DOT) and the International Air Transport Association (IATA) regulations for shipping Biological Substances.
  • Meets DOT and IATA requirements
  • Integrated absorbent sheet
  • Quick tear opening feature
DOT and IATA shipping regulations require that specimens classified as UN3373 Biological Substance, Category B are transported in either a primary receptacle or secondary packaging that has passed the 95 kPa pressure differential test as a part of the triple packaging regulations.
Therapak offers the innovative, award-winning ShuttlePouch for use with kits and packaging that require compliance to IATA Packing Instructions 650 for specimens classified as UN3373 Biological Substance, Category B.
The space-saving ShuttlePouch offers a vast reduction in plastic usage. It is easy to use and has a preinserted absorbent. It is ideal for shipping specimens back to the lab in the original kit packaging or alternate shippers. When ready to use, simply insert the specimen tube, follow the closing instructions printed on the pouch, and complete the kit packing instructions for secure and compliant transport. The pouch features an easy to open tear area to quickly access the specimen tube and facilitate processing when the samples arrive at the lab.
Product Family Options
  • Désignation
    Dimensions
    Ø
    Packaging
  • ShuttlePouch for tube sizes up to 20 mm
    59×190 mm
    110 mm
    250 per case
    Case of 250
    Case of 250 - $627.93
    $627.93
    Each
    Each - $627.93
    $627.93
    Case of 250
       $627.93
    Each
       $627.93
     : CSTGSHU-02-3-2
  • ShuttlePouch for tube sizes up to 20 mm
    59×190 mm
    110 mm
    1000 per case
    Case of 1,000
    Case of 1,000 - $2,025.35
    $2,025.35
    Each
    Each - $2,025.35
    $2,025.35
    Case of 1,000
       $2,025.35
    Each
       $2,025.35
     : CSTGSHU-02-0395KPA
  • ShuttlePouch for tube sizes up to 32 mm
    90×190 mm
    105 mm
    1000 per case
    Case of 1,000
    Case of 1,000 - $2,112.91
    $2,112.91
    Each
    Each - $2,112.91
    $2,112.91
    Case of 1,000
       $2,112.91
    Each
       $2,112.91
     : CSTGSHU-02-0695KPA