Lighthouse Center Pull ES Wipe System Starter Pack
Catalogue # 76716-754
Fournisseur: Unimed-Midwest
Spécifications
- Description:Center pull ES wipe starter pack
- Material:PP
- Cat. no.:76716-754
- W×L:11×11"
Spécifications
À propos de cet article
The Lighthouse Center Pull ES System is engineered and designed to help achieve maximum cleaning results when used in conjunction with optimized technologies to disinfect all surfaces.
- Proprietary textured pattern helps maximize cleaning effectiveness
- Cost-effective solution to daily and discharge surface cleaning
- 30% more cleaning abrasiveness; picks up 11% more soil compared to similar products
- Maintains target disinfection and sanitation concentrations
- 11×11" triple spun polypropylene blend wipe
This single wipe dispensing system prevents cross contamination in any environment. The system boasts 30% more cleaning abrasive power, and can pick up 11% more soil compared to similar products.