Pith Ball Electroscopes | Avantor
Pith Ball Electroscopes
Supplier:   American Scientific
Pith Ball Electroscopes
Certificates
Pith Ball Electroscopes
Pith Ball Electroscopes
2 Options Available Below
Product Details & Documents
Simple, timeless demonstration of electrostatics.
  • Comes with a teacher’s guide
  • Additional pith balls available in packs of 10
Use this item for demonstrating the attraction and repulsion of electric charges. It consists of an insulated brass pillar and hook on a base. It is useful for interesting and engaging demonstrations.
Delivery information: Supplied with two colored pith balls with strings attached.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Pith ball electroscope
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $10.05
    $10.05
    Each
       $10.05
    Supplier Number: 3134-01
  • Pith balls, pack of 10
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $9.05
    $9.05
    Pack of 1
       $9.05
    Supplier Number: 3134-02