Thermo Scientific™ Matrix™ Screw Top Tri-Coded Tubes
Thermo Scientific™ Matrix™ Screw Top Tri-Coded Tubes
Thermo Scientific™ Matrix™ Screw Top Tri-Coded Tubes
Certificates
Thermo Scientific™ Matrix™ Screw Top Tri-Coded Tubes
Thermo Scientific™ Matrix™ Screw Top Tri-Coded Tubes
12 Options Available Below
Product Details & Documents
Securely store precious samples at low temperatures, including vapor phase liquid nitrogen with Thermo Scientific™ Matrix™ 500 µl and 1.0 ml screw top tubes. 
  • Tubes feature a permanent, high contrast 2D,1D linear and human readable barcode.
  • Constructed of virgin class VI medical grade polypropylene
  • ScrewTop caps feature a soft silicone gasket that conforms to mating surfaces to ensure sample preservation
  • Racks are constructed of sturdy acrylonitrile butadiene styrene (ABS) to protect tubes in case of accidental dropping
  • Screw top Tubes are compatible with storage down to –176 °C
Tubes are molded from medical grade resins and feature a permanent, high contrast 2D,1D linear and human readable barcode.
Secure tracking:
Permanent, unique 2D barcode is laser-etched on the bottom of each tube
2D barcode allows you to scan and decode tubes at once without removing them from the rack, and allows data to be associated with each tube
Compatible with our range of VisionMate readers
Superior storage format:
ANSI-format latch racks maximize storage space and are compatible with multichannel manual pipettes or automated liquid handling equipment
Several capping / decapping options available to manipulate a single tube, 8 tubes, or 96 tubes at a time
Stringent quality control:
Every 2D barcode is scanned for readability
Every 2D barcode is checked against the database of all previous Thermo Scientific 2D barcodes to ensure there are zero duplicates
Each tube is leak tested to ensure tube integrity and sample security
All tubes are supplied free from DNA, RNase, DNase, endotoxins and cytotoxins
Certifications: Products are manufactured in ISO 7 class clean room environment according to ISO 14644-1:2015. Sterile products are sterilized by gamma irradiation according to ISO 11137; 10⁻³ Sterility Assurance Level (SAL). The vials and closures were tested and met the requirements of USP <88> Class VI plastics.
Ordering information: These tubes can be ordered to come in racks (barcoded or non-barcoded) or may be purchased in bulk. The tri-coded tubes can be purchased sterile or non-sterile. We also offer tubes with white patch only if your work does not require the 1D linear and human readable code. The 2D screw top tubes meet a range of manual and automated long term storage needs; barcoded racks make it easy to track the rack as well. Use in combination with Thermo Scientific barcode readers, arrayers and capping equipment.
Documents
Product Family Options
Product Information
  • Désignation
    Capacité
    Bottom Style
    Color
    Avec bouchon
    Cap Color
    Sterility
    Packaging
    Availability
    Price
  • 2D barcoded screw top tubes with white patch, 1D linear barcode
    1.00 ml
    V bottom
    Clear
    Oui
    Clear
    Sterile
    Bulk
    Log in for availability
    Log in for availability
    Case of 480
    Case of 480 - $1,174.41
    $1,174.41
    Case of 480
       $1,174.41
    Supplier Number: 3740-WP1D
  • 2D barcoded screw top tubes with white patch, 1D linear barcode, barcoded rack
    0.50 ml
    V bottom
    Clear
    Oui
    Clear
    Non sterile
    5 latch racks of 96 tubes
    Log in for availability
    Log in for availability
    Case of 5
    Case of 5 - $1,500.60
    $1,500.60
    Case of 5
       $1,500.60
    Supplier Number: 3744NS-WP1D-BR
  • 2D barcoded screw top tubes with white patch, 1D linear barcode
    0.50 ml
    V bottom
    Clear
    Non
    Sterile
    5 latch racks of 96 tubes
    Log in for availability
    Log in for availability
    Case of 5
    Case of 5 - $871.19
    $871.19
    Case of 5
       $871.19
    Supplier Number: 3745-WP1D
  • 2D barcoded screw top tubes with white patch, 1D linear barcode
    1.00 ml
    V bottom
    Clear
    Non
    Sterile
    5 latch racks of 96 tubes
    Log in for availability
    Log in for availability
    Case of 5
    Case of 5 - $924.27
    $924.27
    Case of 5
       $924.27
    Minimum order quantity is 1
    Supplier Number: 3742-WP1D
  • 2D barcoded screw top tubes with white patch, 1D linear barcode, barcoded rack
    1.00 ml
    V bottom
    Clear
    Oui
    Clear
    Sterile
    5 latch racks of 96 tubes
    Log in for availability
    Log in for availability
    Case of 5
    Case of 5 - $1,475.78
    $1,475.78
    Case of 5
       $1,475.78
    Supplier Number: 3741-WP1D-BR
  • 2D barcoded screw top tubes with white patch, 1D linear barcode, barcoded rack
    0.50 ml
    V bottom
    Clear
    Non
    Non sterile
    5 latch racks of 96 tubes
    Log in for availability
    Log in for availability
    Case of 5
    Case of 5 - $910.98
    $910.98
    Case of 5
       $910.98
    Supplier Number: 3745NS-WP1D-BR
  • 2D barcoded screw top tubes with white patch, 1D linear barcode
    0.50 ml
    V bottom
    Clear
    Oui
    Clear
    Sterile
    5 latch racks of 96 tubes
    Log in for availability
    Log in for availability
    Case of 5
    Case of 5 - $1,383.08
    $1,383.08
    Case of 5
       $1,383.08
    Supplier Number: 3744-WP1D
  • 2D barcoded screw top tubes with white patch, 1D linear barcode, barcoded rack
    1.00 ml
    V bottom
    Clear
    Non
    Sterile
    5 latch racks of 96 tubes
    Log in for availability
    Log in for availability
    Case of 5
    Case of 5 - $1,008.67
    $1,008.67
    Case of 5
       $1,008.67
    Supplier Number: 3742-WP1D-BR
  • 2D barcoded screw top tubes with white patch, 1D linear barcode
    0.50 ml
    V bottom
    Clear
    Oui
    Clear
    Sterile
    Bulk
    Log in for availability
    Log in for availability
    Case of 480
    Case of 480 - $1,277.08
    $1,277.08
    Case of 480
       $1,277.08
    Supplier Number: 3743-WP1D
  • 2D barcoded screw top tubes with white patch, 1D linear barcode
    1.00 ml
    V bottom
    Clear
    Oui
    Clear
    Sterile
    5 latch racks of 96 tubes
    Log in for availability
    Log in for availability
    Case of 5
    Case of 5 - $1,520.00
    $1,520.00
    Case of 5
       $1,520.00
    Supplier Number: 3741-WP1D
  • 2D barcoded screw top tubes with white patch, 1D linear barcode, barcoded rack
    0.50 ml
    V bottom
    Clear
    Oui
    Clear
    Sterile
    5 latch racks of 96 tubes
    Log in for availability
    Log in for availability
    Case of 5
    Case of 5 - $1,477.44
    $1,477.44
    Case of 5
       $1,477.44
    Supplier Number: 3744-WP1D-BR
  • 2D barcoded screw top tubes with white patch, 1D linear barcode, barcoded rack
    0.50 ml
    V bottom
    Clear
    Non
    Sterile
    5 latch racks of 96 tubes
    Log in for availability
    Log in for availability
    Case of 5
    Case of 5 - $945.76
    $945.76
    Case of 5
       $945.76
    Supplier Number: 3745-WP1D-BR