Thermo Scientific™ Matrix™ Screw Top Tube Caps, Cap Trays and Cap Inserts
Simplify sample storage with these color-coded caps and inserts.
- Gasketed caps ensure critical sample protection over the storage lifetime
- Soft gasket compresses to ensure a proper seal, preventing contamination from outside the tube
- Cap material: Virgin class VI medical grade polypropylene with virgin class VI medical grade silicone O-ring
- ScrewTop caps are compatible with storage down to –196 °C
Thermo Scientific™ Matrix™ ScrewTop Tube caps and inserts are available racked in trays or in bulk to suit a variety of applications.
Cap options simplify sample identification:
Cap trays feature standard microplate format compatible with automated equipment
Caps are available in seven color options to enable sample identification
Color inserts snap into cap for a secure fit; inserts are removable
Inserts are available in six colors
All caps and inserts are supplied free from DNA, Rnase, Dnase, endotoxins and cytotoxins
: Products are manufactured in ISO 7 class clean room environment according to ISO 14644-1:2015.
Sterile products are sterilized by gamma irradiation according to ISO 11137; 10⁻³ Sterility Assurance Level (SAL)
The vials and closures were tested and met the requirements of USP <88> Class VI plastics.
Sterile products are sterilized by gamma irradiation according to ISO 11137; 10⁻³ Sterility Assurance Level (SAL)
The vials and closures were tested and met the requirements of USP <88> Class VI plastics.