Accessories for eZwest™ Lite Automated Western Device
Supplier: Genscript
Certificates
About this item
Frequently Bought Together
VWR® Razor Blades
Supplier: VWR International
Acétone ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
INCYTO C-Chip™ Disposable Hemacytometers, SKC
Supplier: MEDONE SURGICAL, INC
Acétone ≥99.5% (by GC, corrected for water content), OmniSolv®, Supelco®
Supplier: MilliporeSigma
Waste Gas Control Supplies, VetEquip®
Supplier: Vetequip
SOLUTION ELER CLNG 450 ML INORGANIC SUBS
Supplier: Hanna
Éthanol dénaturé, anhydrous
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Corning® RPMI 1640, Corning
Supplier: Corning
VWR® Disposable Petri Dishes
Supplier: VWR International
Single-Use Needles, BD Medical
Supplier: BD
n-Hexane ≥95%, OmniSolv®, Supelco®
Supplier: MilliporeSigma
DAPT ≥98% (by NMR)
Supplier: Enzo Life Sciences
Dri-Contrad® Powder Detergent
Supplier: Decon Labs
MycoAlert™ Mycoplasma Detection Kits, Lonza
Supplier: LONZA PHARMA - BIOSCIENCE CA
Acide acétique glacial ≥99% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Magnesium
Supplier: Ward's Science