Eisco Labs Basic Starter Laboratory Tool Set
Eisco labs basic starter laboratory tool set
Catalog # 470344-258
Supplier: Wards
Fournisseur #: LABTS
Jump to:
Product Details & Documents
Handy set of hand tools ideal for beginners, providing essential tools for basic laboratory work, ensuring convenience and practicality in scientific experiments and educational settings.
- Seven-piece set of essential hand tools
- All items are made of high-quality materials
- Great starter set for students/hobbyists
- All the basics needed for Chemistry labs
Kit includes: (1) Scale (cm one side/inches other side), (1) stainless steel test tube holder, (1) stainless steel spoon (5.75" long), (1) fine point metal tweezers, (1) flask tong, (1) neoprene tube clamp, (1) nylon test tube cleaning bristled brush (4" of brush, approx. 0.75" diameter).
Spécifications
- Désignation:Eisco labs basic starter laboratory tool set