Fractional Distillation Set | Avantor
Fractional Distillation Set
Fractional distillation set
Catalog # 470343-788
Supplier:   Wards
Fournisseur #:  CH1030
Fractional Distillation Set
Catalog # 470343-788
Supplier:  Wards
Fractional Distillation Set
Fractional Distillation Set
Fractional distillation set
Price
$285.69
Log in pour la disponibilité
Each
Product Details & Documents
Comprehensive four-piece set for accurate fractional distillation, including 250 ml distillation flask, Vigrex column, receiver adapter, and still head, all crafted from high-quality borosilicate glass.
  • Fractional distillation process is the separation of a mixture into individual parts of fractions by heating it up
  • Kit includes five pieces: 250 ml round bottom flask, Vigrex column, still head, receiver adaptor
  • All joint sizes are 24/29
  • All pieces are made of high quality, heavy duty borosilicate glass 3.3
  • Perfect for any school or professional laboratory
Spécifications
  • Désignation:
    Fractional distillation set