Simple Distillation Set
Simple distillation set
Catalog # 470345-146
Supplier: Wards
Fournisseur #: CH1026
Jump to:
Product Details & Documents
This four-piece set with 24/29 joints provides the necessary components for simple distillation processes, ensuring efficiency and reliability in laboratory work with its high-quality borosilicate glass construction.
- Simple Distillation set made of high quality Borosilicate glass
- All joint sizes are 24/29
- All pieces are made of high quality, heavy duty Borosilicate Glass 3.3
- Perfect for any school or professional laboratory
Set includes four pieces: 250 ml round bottom flask, still head, Liebig condenser, adaptor receiver.
Spécifications
- Désignation:Simple distillation set